MilliporeSigma
  • Activity of fleroxacin alone and in combination with clindamycin or metronidazole in experimental intra-abdominal abscesses.
Activity of fleroxacin alone and in combination with clindamycin or metronidazole in experimental intra-abdominal abscesses.
Antimicrobial agents and chemotherapy (1994-02-01)
A Pefanis, C Thauvin-Eliopoulos, J Holden, G M Eliopoulos, M J Ferraro, R C Moellering
ABSTRACT
To assess the potential efficacy of fleroxacin in combination with clindamycin or metronidazole in mixed aerobic and anaerobic infections, we used a rat model of intra-abdominal abscesses in which the inoculum consisted of pooled rat feces mixed with BaSO4. Two hours after bacterial challenge, antimicrobial therapy was begun intravenously with regimens designed to stimulate human pharmacokinetics. A combination of clindamycin and gentamicin was included as an established treatment regimen. After 8.5 days of therapy, final bacterial counts in abscesses showed that fleroxacin alone or combined with metronidazole or clindamycin effectively eradicated Escherichia coli, with bacterial densities of < or = 2.84 +/- 0.1, < or = 2.9 +/- 0.1, and < or = 2.9 +/- 0.1 (mean +/- standard error of the mean) log10 CFU/g, respectively. The addition of either clindamycin or metronidazole to fleroxacin substantially enhanced the effectiveness of the regimens against Bacteroides fragilis, with bacterial counts of < or = 3.0 +/- 0.1 or < or = 2.9 +/- 0.1 log10 CFU/g, respectively, versus 9.2 +/- 0.2 log10 CFU/g for fleroxacin alone. The combination of metronidazole and fleroxacin also resulted in a significantly greater reduction of peptostreptococci and Bacteroides thetaiotaomicron than fleroxacin alone (< or = 2.9 +/- 0.1 versus 6.1 +/- 0.9 log10 CFU/g and 3.3 +/- 0.4 versus 8.3 +/- 0.1 log10 CFU/g, respectively). Except for those of B. fragilis, counts of other anaerobes were reduced to a greater extent by metronidazole plus fleroxacin than by clindamycin plus fleroxacin, although differences were not always significant. Metronidazole plus fleroxacin was at least as active a clindamycin plus gentamicin against all species and was significantly more active against Clostridium spp. No regimen effectively eradicated enterococci from the abscesses. These results suggest that the addition of either metronidazole or clindamycin would effectively enhance the spectrum of fleroxacin for treatment of mixed aerobic and anaerobic infections.