MilliporeSigma
  • High-performance liquid chromatographic determination of N1-alkylnicotinamide in urine.
High-performance liquid chromatographic determination of N1-alkylnicotinamide in urine.
Analytical biochemistry (1985-05-15)
T Hirayama, K Yoshida, K Uda, M Nohara, S Fukui
ABSTRACT
A simple high-performance liquid chromatographic method has been developed for determining N1-alkylnicotinamides, including C1-C5 alkyl derivatives, in urine. N1-Alkylnicotinamides were reacted with acetophenone in strong alkali medium at 0 degrees C and then formic acid was added. The reaction mixture was heated in acidic medium at above 93 degrees C, and the fluorescent product, 1-alkyl-7-phenyl-1,5-dihydro-5-oxo-1,6-naphthyridine, was chromatographed by HPLC, using a Zorbax SCX-300 column with a mixed mobile phase of acetonitrile-0.04 M ammonium phosphate, monobasic. N1-Alkylnicotinamides can be determined as 1,6-naphthyridine derivatives by a fluorometric detector at a level of 100 pg (signal/noise = 2). Recoveries of N1-alkylnicotinamides in urine were satisfactory. Interfering reaction products from NAD+ and NADP+ were clearly eliminated for determination of N1-alkylnicotinamides without pentyl derivatives.
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)