MilliporeSigma
The inositol trisphosphate (IP3) signal transduction pathway.
Science signaling (2010-04-29)
Jack D Thatcher
ABSTRACT
This resource provides and describes two animated lessons that illustrate inositol 1,4,5-trisphosphate signal transduction pathways. They are designed for any collegiate-level course that presents the basic steps of the prototypical pathway. Courses that might employ these animations include Introductory Biology, Cell Biology, Physiology, Biochemistry, Molecular Biology, and Pharmacology.
Product Description
Avanti
IP3(1,4,5), Avanti Research - A Croda Brand