MilliporeSigma
  • Coupled assay of sphingomyelin and ceramide molecular species by gas liquid chromatography.
Coupled assay of sphingomyelin and ceramide molecular species by gas liquid chromatography.
Journal of lipid research (2002-03-15)
Claude Vieu, François Tercé, Françoise Chevy, Corinne Rolland, Ronald Barbaras, Hugues Chap, Claude Wolf, Bertrand Perret, Xavier Collet
ABSTRACT
This study reports a single-step analysis of the molecular species of endogenous ceramides and of the ceramide moiety of sphingomyelins in biological samples, using gas liquid chromatography (GLC). Silylated sphingomyelins were quantitatively converted to monosilylated ceramide upon injection into GLC, whereas the free ceramides were di-silylated on the primary and secondary alcohol function, as confirmed by mass spectrometry. The reproducible shift of the retention times between the mono- and di-silylated derivatives enables simultaneous quantification of the variety of sphingomyelin and ceramide molecular species. Overlapping diacylglycerols were first removed by a mild alkaline treatment of the lipid extract. The lowest detection limit (5 pmol) did not allow for identification of free ceramides in human plasma, but 17 molecular species of ceramides derived from sphingomyelins were quantified, from NC16:0 up to NC24:1. By contrast, three major free ceramides (NC16:0, NC24:0, and NC24:1) were quantified in HEPG2 and Chinese hamster ovary (CHO) cells. Upon induction of apoptosis in CHO cells by C6-ceramide, we could follow the disappearance of the C6-ceramide, its partial conversion to C6-sphingomyelin, and the prominent increase of NC16:0 ceramide. Thus, our method represents a unique procedure of simultaneous analysis of sphingomyelin and ceramide molecular species able to monitor the variation of the different pools in biological samples.
Product Description
Avanti
C15 Ceramide-d7 (d18:1-d7/15:0), Avanti Research - A Croda Brand 860681P, powder