MilliporeSigma
  • Quantification of ceramide species in biological samples by liquid chromatography electrospray ionization tandem mass spectrometry.
Quantification of ceramide species in biological samples by liquid chromatography electrospray ionization tandem mass spectrometry.
Analytical biochemistry (2010-02-25)
Takhar Kasumov, Hazel Huang, Yoon-Mi Chung, Renliang Zhang, Arthur J McCullough, John P Kirwan
ABSTRACT
We present an optimized and validated liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the simultaneous measurement of concentrations of different ceramide species in biological samples. The method of analysis of tissue samples is based on Bligh and Dyer extraction, reverse-phase high-performance liquid chromatography separation, and multiple reaction monitoring of ceramides. Preparation of plasma samples also requires isolation of sphingolipids by silica gel column chromatography prior to LC-ESI-MS/MS analysis. The limits of quantification were in a range of 0.01-0.50ng/ml for distinct ceramides. The method was reliable for inter- and intraassay precision, accuracy, and linearity. Recoveries of ceramide subspecies from human plasma, rat liver, and muscle tissue were 78 to 91%, 70 to 99%, and 71 to 95%, respectively. The separation and quantification of several endogenous long-chain and very-long-chain ceramides using two nonphysiological odd chain ceramide (C17 and C25) internal standards was achieved within a single 21-min chromatographic run. The technique was applied to quantify distinct ceramide species in different rat tissues (muscle, liver, and heart) and in human plasma. Using this analytical technique, we demonstrated that a clinical exercise training intervention reduces the levels of ceramides in plasma of obese adults. This technique could be extended for quantification of other ceramides and sphingolipids with no significant modification.
Product Description
Avanti
C18 Ceramide-d7 (d18:1-d7/18:0), Avanti Research - A Croda Brand 860677P, powder