MilliporeSigma
  • Separation of cis-trans phospholipid isomers using reversed phase LC with high resolution MS detection.
Separation of cis-trans phospholipid isomers using reversed phase LC with high resolution MS detection.
Analytical chemistry (2012-06-05)
Susan S Bird, Vasant R Marur, Irina G Stavrovskaya, Bruce S Kristal
ABSTRACT
The increased presence of synthetic trans fatty acids into western diets has been shown to have deleterious effects on physiology and raising an individual's risk of developing metabolic disease, cardiovascular disease, and stroke. The importance of these fatty acids for health and the diversity of their (patho) physiological effects suggest that not only should the free trans fatty acids be studied but also monitoring the presence of these fats into the side chains of biological lipids, such as glycerophospholipids, is also essential. We developed a high resolution LC-MS method that quantitatively monitors the major lipid classes found in biospecimens in an efficient, sensitive, and robust manner while also characterizing individual lipid side chains through the use of high energy collisional dissociation (HCD) fragmentation and chromatographic alignment. We herein show how this previously described reversed phase method can baseline separate the cis-trans isomers of phosphatidylglycerol and phosphatidylcholine (PC) with two 18:1 side chains, in both positive and negative mode, as neat solutions and when spiked into a biological matrix. Endogenous PC (18:1/18:1)-cis and PC (18:1/18:1)-trans isomers were examined in mitochondrial and serum profiling studies, where rats were fed diets enriched in either trans 18:1 fatty acids or cis 18:1 fatty acids. In this study, we determined the cis:trans isomer ratios of PC (18:1/18:1) and related this ratio to dietary composition. This generalized LC-MS method enables the monitoring of trans fats in biological lipids in the context of a nontargeted method, allowing for relative quantitation and enhanced identification of unknown lipids in complex matrixes.
Product Description
Avanti
18:0-18:2 PG, 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder
Avanti
18:1 (Δ9-Trans) PG, 1,2-dielaidoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder
Avanti
18:0-18:2 PG, 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform