MilliporeSigma
  • Protein transport across membranes: Comparison between lysine and guanidinium-rich carriers.
Protein transport across membranes: Comparison between lysine and guanidinium-rich carriers.
Biochimica et biophysica acta (2015-09-08)
Max Lein, Brittany M deRonde, Federica Sgolastra, Gregory N Tew, Matthew A Holden
ABSTRACT
The mechanism(s) by which certain small peptides and peptide mimics carry large cargoes across membranes through exclusively non-covalent interactions has been difficult to resolve. Here, we use the droplet-interface bilayer as a platform to characterize distinct mechanistic differences between two such carriers: Pep-1 and a guanidinium-rich peptide mimic we call D9. While both Pep-1 and D9 can carry an enzyme, horseradish peroxidase (HRP) across a lipid bilayer, we found that they do so by different mechanisms. Specifically, Pep-1 requires voltage or membrane asymmetry while D9 does not. In addition, D9 can facilitate HRP transport without pre-forming a complex with HRP. By contrast, complex formation is required by Pep-1. Both carriers are capable of forming pores in membranes but our data hints that these pores are not responsible for cargo transport. Overall, D9 appears to be a more potent and versatile transporter when compared with Pep-1 because D9 does not require an applied voltage or other forces to drive transport. Thus, D9 might be used to deliver cargo across membranes under conditions where Pep-1 would be ineffective.
Product Description
Avanti
4ME 16:0 PG, 1,2-diphytanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder
Avanti
4ME 16:0 PG, 1,2-diphytanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform