MilliporeSigma
  • Physical determinants of β-barrel membrane protein folding in lipid vesicles.
Physical determinants of β-barrel membrane protein folding in lipid vesicles.
Biophysical journal (2011-05-05)
Alison H Dewald, Jacqueline C Hodges, Linda Columbus
ABSTRACT
The spontaneous folding of two Neisseria outer membrane proteins, opacity-associated (Opa)(60) and Opa(50) into lipid vesicles was investigated by systematically varying bulk and membrane properties. Centrifugal fractionation coupled with sodium dodecyl sulfate polyacrylamide gel electrophoresis mobility assays enabled the discrimination of aggregate, unfolded membrane-associated, and folded membrane-inserted protein states as well as the influence of pH, ionic strength, membrane surface potential, lipid saturation, and urea on each. Protein aggregation was reduced with increasing lipid chain length, basic pH, low salt, the incorporation of negatively charged guest lipids, or by the addition of urea to the folding reaction. Insertion from the membrane-associated form was improved in shorter chain lipids, with more basic pH and low ionic strength; it is hindered by unsaturated or ether-linked lipids. The isolation of the physical determinants of insertion suggests that the membrane surface and dipole potentials are driving forces for outer membrane protein insertion and folding into lipid bilayers.
Product Description
Avanti
10:0 PG, 1,2-didecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder