MilliporeSigma
  • Systems Analyses Reveal Physiological Roles and Genetic Regulators of Liver Lipid Species.
Systems Analyses Reveal Physiological Roles and Genetic Regulators of Liver Lipid Species.
Cell systems (2018-06-18)
Pooja Jha, Molly T McDevitt, Rahul Gupta, Pedro M Quiros, Evan G Williams, Karim Gariani, Maroun B Sleiman, Leo Diserens, Adam Jochem, Arne Ulbrich, Joshua J Coon, Johan Auwerx, David J Pagliarini
ABSTRACT
The genetics of individual lipid species and their relevance in disease is largely unresolved. We profiled a subset of storage, signaling, membrane, and mitochondrial liver lipids across 385 mice from 47 strains of the BXD mouse population fed chow or high-fat diet and integrated these data with complementary multi-omics datasets. We identified several lipid species and lipid clusters with specific phenotypic and molecular signatures and, in particular, cardiolipin species with signatures of healthy and fatty liver. Genetic analyses revealed quantitative trait loci for 68% of the lipids (lQTL). By multi-layered omics analyses, we show the reliability of lQTLs to uncover candidate genes that can regulate the levels of lipid species. Additionally, we identified lQTLs that mapped to genes associated with abnormal lipid metabolism in human GWASs. This work provides a foundation and resource for understanding the genetic regulation and physiological significance of lipid species.
Product Description
Avanti
15:0 PC, 1,2-dipentadecanoyl-sn-glycero-3-phosphocholine, powder
Avanti
17:0 PA, 1,2-diheptadecanoyl-sn-glycero-3-phosphate (sodium salt), powder
Avanti
17:0 PS, Avanti Research - A Croda Brand