MilliporeSigma
  • Delivery of a drug cache to glioma cells overexpressing platelet-derived growth factor receptor using lipid nanocarriers.
Delivery of a drug cache to glioma cells overexpressing platelet-derived growth factor receptor using lipid nanocarriers.
Nanomedicine (London, England) (2016-03-24)
Kayla Miller, Suraj Dixit, Amy-Lee Bredlau, Alfred Moore, Emilie McKinnon, Ann-Marie Broome
ABSTRACT
Glioblastoma multiforme is a devastating disease with no curative options due to the difficulty in achieving sufficient quantities of effective chemotherapies into the tumor past the blood-brain barrier. Micelles loaded with temozolomide (TMZ) were designed to increase the delivery of this drug into the brain. pH-responsive micelles composed of distearoyl phosphoethanolamine-PEG-2000-amine and N-palmitoyl homocysteine were surface-functionalized with PDGF peptide and Dylight 680 fluorophore. PDGF-micelles containing TMZ have specific uptake and increased killing in glial cells compared with untargeted micelles. In vivo studies demonstrated selective accumulation of PDGF-micelles containing TMZ in orthotopic gliomas implanted in mice. Targeted micelle-based drug carrier systems hold potential for delivery of a wide variety of hydrophobic drugs thereby reducing its systemic toxicity.
Product Description
Avanti
N-palmitoyl homocysteine (PHC), Avanti Research - A Croda Brand 870320P, powder