MilliporeSigma
  • Liquiritigenin is a plant-derived highly selective estrogen receptor beta agonist.
Liquiritigenin is a plant-derived highly selective estrogen receptor beta agonist.
Molecular and cellular endocrinology (2008-01-08)
Jennifer E Mersereau, Nitzan Levy, Richard E Staub, Scott Baggett, Tatjana Zogovic, Tetjana Zogric, Sylvia Chow, William A Ricke, Mary Tagliaferri, Isaac Cohen, Leonard F Bjeldanes, Dale C Leitman
ABSTRACT
After the Women's Health Initiative found that the risks of hormone therapy outweighed the benefits, a need for alternative drugs to treat menopausal symptoms has emerged. We explored the possibility that botanical agents used in Traditional Chinese Medicine for menopausal symptoms contain ERbeta-selective estrogens. We previously reported that an extract containing 22 herbs, MF101 has ERbeta-selective properties. In this study we isolated liquiritigenin, the most active estrogenic compound from the root of Glycyrrhizae uralensis Fisch, which is one of the plants found in MF101. Liquiritigenin activated multiple ER regulatory elements and native target genes with ERbeta but not ERalpha. The ERbeta-selectivity of liquiritigenin was due to the selective recruitment of the coactivator steroid receptor coactivator-2 to target genes. In a mouse xenograph model, liquiritigenin did not stimulate uterine size or tumorigenesis of MCF-7 breast cancer cells. Our results demonstrate that some plants contain highly selective estrogens for ERbeta.
Sigma-Aldrich
Liquiritigenin, ≥97.0% (HPLC)