MilliporeSigma
  • Natural cardenolides suppress coronaviral replication by downregulating JAK1 via a Na+/K+-ATPase independent proteolysis.
Natural cardenolides suppress coronaviral replication by downregulating JAK1 via a Na+/K+-ATPase independent proteolysis.
Biochemical pharmacology (2020-06-28)
Cheng-Wei Yang, Hsing-Yu Hsu, Hsin-Yu Chang, Yue-Zhi Lee, Shiow-Ju Lee
ABSTRACT
An unprecedented biological function of natural cardenolides independent of their membrane target Na+/K+-ATPase is disclosed. Previously, we reported that cardenolides impart anti-transmissible gastroenteritis coronavirus (anti-TGEV) activity through the targeting of Na+/K+-ATPase and its associated PI3K_PDK1_RSK2 signaling. Swine testis cells with Na+/K+-ATPase α1 knocked down exhibited decreased susceptibility to TGEV infectivity and attenuated PI3K_PDK1_RSK2 signaling. Herein, we further explored a Na+/K+-ATPase-independent signaling axis induced by natural cardenolides that also afforded significant anti-coronaviral activity for porcine TGEV and human HCoV-OC43. Using pharmacological inhibition and gene silencing techniques, we found that this anti-TGEV or anti-HCoV-OC43 activity was caused by JAK1 proteolysis and mediated through upstream activation of Ndfip1/2 and its effector NEDD4. This study provides novel insights into the pharmacological effects of natural cardenolides, and is expected to inform their future development as antiviral agents.
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Digitoxigenin
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder