MilliporeSigma
  • A central role of IKK2 and TPL2 in JNK activation and viral B-cell transformation.
A central role of IKK2 and TPL2 in JNK activation and viral B-cell transformation.
Nature communications (2020-02-06)
Stefanie Voigt, Kai R Sterz, Fabian Giehler, Anne-Wiebe Mohr, Joanna B Wilson, Andreas Moosmann, Arnd Kieser
ABSTRACT
IκB kinase 2 (IKK2) is well known for its pivotal role as a mediator of the canonical NF-κB pathway, which has important functions in inflammation and immunity, but also in cancer. Here we identify a novel and critical function of IKK2 and its co-factor NEMO in the activation of oncogenic c-Jun N-terminal kinase (JNK) signaling, induced by the latent membrane protein 1 (LMP1) of Epstein-Barr virus (EBV). Independent of its kinase activity, the TGFβ-activated kinase 1 (TAK1) mediates LMP1 signaling complex formation, NEMO ubiquitination and subsequent IKK2 activation. The tumor progression locus 2 (TPL2) kinase is induced by LMP1 via IKK2 and transmits JNK activation signals downstream of IKK2. The IKK2-TPL2-JNK axis is specific for LMP1 and differs from TNFα, Interleukin-1 and CD40 signaling. This pathway mediates essential LMP1 survival signals in EBV-transformed human B cells and post-transplant lymphoma, and thus qualifies as a target for treatment of EBV-induced cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human IKBKB
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP3K8
Sigma-Aldrich
Monoclonal Anti-MYC , (C-terminal) antibody produced in mouse, clone 9E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-FLAG® antibody, Rat monoclonal, clone 6F7, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human CHUK