MilliporeSigma
  • Distinct clinical and prognostic implication of IDH1/2 mutation and other most frequent mutations in large duct and small duct subtypes of intrahepatic cholangiocarcinoma.
Distinct clinical and prognostic implication of IDH1/2 mutation and other most frequent mutations in large duct and small duct subtypes of intrahepatic cholangiocarcinoma.
BMC cancer (2020-04-16)
Bingqi Ma, Huijuan Meng, Ye Tian, Yingying Wang, Tianqiang Song, Ti Zhang, Qiang Wu, Yunlong Cui, Huikai Li, Wei Zhang, Qiang Li
ABSTRACT
Isocitrate dehydrogenase 1/2 (IDH1/2), BAP1, ARID1A and PBRM1 have been reported as the most frequent mutant genes in intrahepatic cholangiocarcinoma (ICC), and their relationships with clinicopathological features and prognosis were researched in this study. We collected clinical data of 130 ICC patients from January 2012 to December 2017. The IDH1/2 mutation and loss of BAP1, ARID1A and PBRM1 expressions were detected by DNA sequencing or immunohistochemical methods, and histological subtype of ICCs was determined by hematoxylin-eosin, Alcian blue and S100P staining. IDH1/2 mutation was related to decreased preoperative serum total bilirubin (P = 0.039), ferritin (P = 0.000) and higher histological differentiation (P = 0.024), and was associated with prolonged disease-free survival (P = 0.009) and a trend toward increased overall survival (P = 0.126) in small duct type of ICCs. Immunohistochemical staining results of MsMab-1 were generally consistent with DNA sequencing for IDH1/2 mutant in ICCs (κ = 0.691). Only BAP1 expression loss was correlated to prolonged disease-free survival (P = 0.031) and overall survival (P = 0.041) in large duct type of ICCs. IDH1/2 mutation is a favorable predictor and may be related to iron metabolism in small duct type of ICCs. Furthermore, we suggest that the detection of IDH1/2 mutation is indispensable to determine targeted therapy in small duct type ICCs, while it is not necessary in large duct of ICCs. MsMab-1 is a relatively effective multi-specific antibody against IDH1/2 mutant in ICCs. BAP1 expression loss was correlated with improved prognosis only in large duct type ICCs.
Sigma-Aldrich
Anti-PBRM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution