MilliporeSigma
IRE1α Promotes Zika Virus Infection via XBP1.
Viruses (2020-03-07)
Elena P Kolpikova, Ana R Tronco, Andreas B den Hartigh, Konner J Jackson, Takao Iwawaki, Susan L Fink
ABSTRACT
Zika virus (ZIKV) is an emergent member of the Flaviviridae family which causes severe congenital defects and other major sequelae, but the cellular processes that support ZIKV replication are incompletely understood. Related flaviviruses use the endoplasmic reticulum (ER) as a membranous platform for viral replication and induce ER stress during infection. Our data suggest that ZIKV activates IRE1α, a component of the cellular response to ER stress. IRE1α is an ER-resident transmembrane protein that possesses a cytosolic RNase domain. Upon activation, IRE1α initiates nonconventional cytoplasmic splicing of XBP1 mRNA. Spliced XBP1 encodes a transcription factor, which upregulates ER-related targets. We find that ZIKV infection induces XBP1 mRNA splicing and induction of XBP1 target genes. Small molecule inhibitors of IRE1α, including those specific for the nuclease function, prevent ZIKV-induced cytotoxicity, as does genetic disruption of IRE1α. Optimal ZIKV RNA replication requires both IRE1α and XBP1. Spliced XBP1 has been described to cause ER expansion and remodeling and we find that ER redistribution during ZIKV infection requires IRE1α nuclease activity. Finally, we demonstrate that inducible genetic disruption of IRE1α and XBP1 impairs ZIKV replication in a mouse model of infection. Together, our data indicate that the ER stress response component IRE1α promotes ZIKV infection via XBP1 and may represent a potential therapeutic target.
Sigma-Aldrich
Monoclonal Anti-PDIA3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2444, purified immunoglobulin, buffered aqueous glycerol solution