MilliporeSigma
  • tappAS: a comprehensive computational framework for the analysis of the functional impact of differential splicing.
tappAS: a comprehensive computational framework for the analysis of the functional impact of differential splicing.
Genome biology (2020-05-20)
Lorena de la Fuente, Ángeles Arzalluz-Luque, Manuel Tardáguila, Héctor Del Risco, Cristina Martí, Sonia Tarazona, Pedro Salguero, Raymond Scott, Alberto Lerma, Ana Alastrue-Agudo, Pablo Bonilla, Jeremy R B Newman, Shunichi Kosugi, Lauren M McIntyre, Victoria Moreno-Manzano, Ana Conesa
ABSTRACT
Recent advances in long-read sequencing solve inaccuracies in alternative transcript identification of full-length transcripts in short-read RNA-Seq data, which encourages the development of methods for isoform-centered functional analysis. Here, we present tappAS, the first framework to enable a comprehensive Functional Iso-Transcriptomics (FIT) analysis, which is effective at revealing the functional impact of context-specific post-transcriptional regulation. tappAS uses isoform-resolved annotation of coding and non-coding functional domains, motifs, and sites, in combination with novel analysis methods to interrogate different aspects of the functional readout of transcript variants and isoform regulation. tappAS software and documentation are available at https://app.tappas.org.
Sigma-Aldrich
Anti-p120 Catenin Antibody, clone 15D2, clone 15D2, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit