MilliporeSigma
  • Evaluation of Glycosylated PTGS2 in Colorectal Cancer for NSAIDS-Based Adjuvant Therapy.
Evaluation of Glycosylated PTGS2 in Colorectal Cancer for NSAIDS-Based Adjuvant Therapy.
Cells (2020-03-15)
Roberta Venè, Delfina Costa, Raffaella Augugliaro, Sebastiano Carlone, Stefano Scabini, Gianmaria Casoni Pattacini, Maurizio Boggio, Simonetta Zupo, Federica Grillo, Luca Mastracci, Francesca Pitto, Simona Minghelli, Nicoletta Ferrari, Francesca Tosetti, Emanuele Romairone, Maria C Mingari, Alessandro Poggi, Roberto Benelli
ABSTRACT
Observational/retrospective studies indicate that prostaglandin-endoperoxide synthase-2 (PTGS2) inhibitors could positively affect colorectal cancer (CRC) patients' survival after diagnosis. To obtain an acceptable cost/benefit balance, the inclusion of PTGS2 inhibitors in the adjuvant setting needs a selective criterion. We quantified the 72 kDa, CRC-associated, glycosylated form of PTGS2 in 100 frozen CRC specimens and evaluated PTGS2 localization by IHC in the same tumors, scoring tumor epithelial-derived and stroma-derived fractions. We also investigated the involvement of interleukin-1 beta (IL1β) in PTGS2 induction, both in vitro and in CRC lysates. Finally, we used overall survival (OS) as a criterion for patient selection. Glycosylated PTGS2 can be quantified with high sensibility in tissue lysates, but the expression in both tumor and stromal cells limits its use for predictive purposes. Immunohistochemistry (IHC) analysis indicates that stromal PTGS2 expression could exert a protective role on patient OS. Stromal PTGS2 was prevalently expressed by cancer-associated fibroblasts exerting a barrier function near the gut lumen, and it apparently favored the antitumor M1 macrophage population. IL1β was directly linked to gPTGS2 expression both in vitro and in tumors, but its activity was apparently prevalent on the stromal cell population. We suggest that stromal PTGS2 could exert a positive effect on patients OS when expressed in the luminal area of the tumor.
Sigma-Aldrich
Monoclonal Anti-MRC1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0387, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution