MilliporeSigma
  • FUT1 deficiency elicits immune dysregulation and corneal opacity in steady state and under stress.
FUT1 deficiency elicits immune dysregulation and corneal opacity in steady state and under stress.
Cell death & disease (2020-04-26)
Kyoung Woo Kim, Jin Suk Ryu, Jung Hwa Ko, Jun Yeob Kim, Hyeon Ji Kim, Hyun Ju Lee, Jang-Hee Oh, Jin Ho Chung, Joo Youn Oh
ABSTRACT
Fucosylation is a biological process that plays a critical role in multiple cellular functions from cell adhesion to immune regulation. Fucosyltransferases (FUTs) mediate fucosylation, and dysregulation of genes encoding FUTs is associated with various diseases. FUT1 and its fucosylated products are expressed in the ocular surface and ocular adnexa; however, the role of FUT1 in the ocular surface health and disease is yet unclear. Here, we investigated the effects of FUT1 on the ocular surface in steady-state conditions with age and under desiccating stress using a Fut1 knockout (KO) mouse model. We found that corneal epithelial defects and stromal opacity developed in Fut1 KO mice. Also, inflammatory responses in the ocular surface and Th1 cell activation in ocular draining lymph nodes (DLNs) were upregulated. Desiccating stress further aggravated Th1 cell-mediated immune responses in DLNs, lacrimal gland, and ocular surface in Fut1 KO mice, leading to severe corneal epithelial disruption and opacity. Mixed lymphocyte reaction assays revealed that the activity of splenocytes to stimulate CD4 T-cell proliferation was increased in Fut1 KO mice. Together, these data demonstrate that FUT1 deficiency induces immune dysregulation in the ocular surface and corneal opacity in steady state and under desiccating stress.
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, Rhodamine conjugate, Chemicon®, from goat