MilliporeSigma
  • Uncoupling of invasive bacterial mucosal immunogenicity from pathogenicity.
Uncoupling of invasive bacterial mucosal immunogenicity from pathogenicity.
Nature communications (2020-04-26)
Simona P Pfister, Olivier P Schären, Luca Beldi, Andrea Printz, Matheus D Notter, Mohana Mukherjee, Hai Li, Julien P Limenitakis, Joel P Werren, Disha Tandon, Miguelangel Cuenca, Stefanie Hagemann, Stephanie S Uster, Miguel A Terrazos, Mercedes Gomez de Agüero, Christian M Schürch, Fernanda M Coelho, Roy Curtiss, Emma Slack, Maria L Balmer, Siegfried Hapfelmeier
ABSTRACT
There is the notion that infection with a virulent intestinal pathogen induces generally stronger mucosal adaptive immunity than the exposure to an avirulent strain. Whether the associated mucosal inflammation is important or redundant for effective induction of immunity is, however, still unclear. Here we use a model of auxotrophic Salmonella infection in germ-free mice to show that live bacterial virulence factor-driven immunogenicity can be uncoupled from inflammatory pathogenicity. Although live auxotrophic Salmonella no longer causes inflammation, its mucosal virulence factors remain the main drivers of protective mucosal immunity; virulence factor-deficient, like killed, bacteria show reduced efficacy. Assessing the involvement of innate pathogen sensing mechanisms, we show MYD88/TRIF, Caspase-1/Caspase-11 inflammasome, and NOD1/NOD2 nodosome signaling to be individually redundant. In colonized animals we show that microbiota metabolite cross-feeding may recover intestinal luminal colonization but not pathogenicity. Consequent immunoglobulin A immunity and microbial niche competition synergistically protect against Salmonella wild-type infection.
Sigma-Aldrich
Anti-Mouse IgA (α-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution