MilliporeSigma
  • R-spondins engage heparan sulfate proteoglycans to potentiate WNT signaling.
R-spondins engage heparan sulfate proteoglycans to potentiate WNT signaling.
eLife (2020-05-21)
Ramin Dubey, Peter van Kerkhof, Ingrid Jordens, Tomas Malinauskas, Ganesh V Pusapati, Joseph K McKenna, Dan Li, Jan E Carette, Mitchell Ho, Christian Siebold, Madelon Maurice, Andres M Lebensohn, Rajat Rohatgi
ABSTRACT
R-spondins (RSPOs) amplify WNT signaling during development and regenerative responses. We previously demonstrated that RSPOs 2 and 3 potentiate WNT/β-catenin signaling in cells lacking leucine-rich repeat-containing G-protein coupled receptors (LGRs) 4, 5 and 6 (Lebensohn and Rohatgi, 2018). We now show that heparan sulfate proteoglycans (HSPGs) act as alternative co-receptors for RSPO3 using a combination of ligand mutagenesis and ligand engineering. Mutations in RSPO3 residues predicted to contact HSPGs impair its signaling capacity. Conversely, the HSPG-binding domains of RSPO3 can be entirely replaced with an antibody that recognizes heparan sulfate (HS) chains attached to multiple HSPGs without diminishing WNT-potentiating activity in cultured cells and intestinal organoids. A genome-wide screen for mediators of RSPO3 signaling in cells lacking LGRs 4, 5 and 6 failed to reveal other receptors. We conclude that HSPGs are RSPO co-receptors that potentiate WNT signaling in the presence and absence of LGRs.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Goat Anti-Human Ig κ chain Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from goat
Sigma-Aldrich
Valproic acid sodium salt, 98%