MilliporeSigma
  • Central galanin receptor 2 mediates galanin action to promote systemic glucose metabolism of type 2 diabetic rats.
Central galanin receptor 2 mediates galanin action to promote systemic glucose metabolism of type 2 diabetic rats.
Biochemical pharmacology (2018-09-01)
Penghua Fang, Biao He, Mei Yu, Mingyi Shi, Yan Zhu, Zhenwen Zhang, Ping Bo
ABSTRACT
Although recent results of our and other studies have showed that galanin (GAL) is an antidiabetic and anti-inflammatory neuropeptide, the molecular mechanism how central GAL regulates energy homeostasis and insulin sensitivity is still not fully understood. The aim of this study was to investigate whether central type 2 of GAL receptors (GALR2) are involved in the regulation of systemic glucose metabolism and its underlying mechanisms. In the present study, type 2 diabetic rats were intracerebroventricularly (i.c.v.) given 100 nM/kg/d GALR2 agonist M1145 or GALR2 antagonist M871 in 5 μl artificial cerebrospinal fluid once a day for consecutive 21 days. Then insulin resistance indexes, inflammatory factor and many genes associated with the function of glucose metabolism were examined in peripheral tissues. The present findings showed that the intracerebroventricular injection of M1145 or M871 respectively increased or decreased glucose infusion rates in hyperinsulinemic euglycemic clamp tests, but attenuated or enhanced the plasma inflammatory factors and glucose concentration in type 2 diabetic rats. Moreover, administration of M1145 markedly increased PGC-1α and GLUT4 expression in skeletal muscles and adipocytes of type 2 diabetic rats. In conclusion, activation of central GALR2 promotes glucose metabolism and ameliorates insulin resistance mainly through the PGC-1α/GLUT4 pathways. The central GALR2 is crucial to whole-body insulin sensitivity and energy homeostasis.
Sigma-Aldrich
M871 trifluoroacetate, ≥95% (HPLC)