MilliporeSigma
  • Comparative exome sequencing reveals novel candidate genes for retinitis pigmentosa.
Comparative exome sequencing reveals novel candidate genes for retinitis pigmentosa.
EBioMedicine (2020-05-27)
Zhen Yi, Jiamin Ouyang, Wenmin Sun, Shiqiang Li, Xueshan Xiao, Qingjiong Zhang
ABSTRACT
Retinitis pigmentosa (RP) is the most common form of inherited retinal degeneration, but genetic defects in nearly half of families remain unresolved. This study aims to identify novel genes associated with RP based on whole exome sequencing (WES) data from 552 probands with RP. Biallelic loss-of-function (LoF) variants were selected from the WES data of 552 probands with RP and compared with that of 4728 in-house controls and the gnomAD database. Expression analysis and knockout mice model or knockdown zebrafish model were performed to confirm the association of a few candidate genes with RP. Unique biallelic LoF variants in ENSA, DACT2, DDR1, and CCDC188 were identified in four probands with RP, but were absent in 4728 in-house controls and were extremely rare in the gnomAD database. The expression of ENSA was enriched in the rod outer segments of human retina, and significant reduced responses of rods and cones were detected in Ensa knockout mice compared to wild-type mice by electroretinogram. The DACT2 transcript showed the highest expression in human retina and knockdown of dact2 in zebrafish resulted in photoreceptor disc membrane disarrangement. This study suggests that ENSA is likely a novel gene for RP and DACT2 is a potentially candidate gene for RP. Further studies are expected to evaluate the association between mutations in the other two genes and RP. To our knowledge, mutations in these genes have not been reported to be associated with RP before.
Product Description
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-DACT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1