MilliporeSigma
  • Vibrational spectroscopic (FTIR and FT Raman) studies, first order hyperpolarizabilities and HOMO, LUMO analysis of p-toluenesulfonyl isocyanate using ab initio HF and DFT methods.
Vibrational spectroscopic (FTIR and FT Raman) studies, first order hyperpolarizabilities and HOMO, LUMO analysis of p-toluenesulfonyl isocyanate using ab initio HF and DFT methods.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-07-29)
K Parimala, V Balachandran
ABSTRACT
The Fourier transform infrared (FTIR) and FT Raman spectra of p-toluenesulfonyl isocyanate (p-tosyl isocyanate) have been measured. The molecular geometry, vibrational frequencies, infrared intensities, Raman activities and atomic charges have been calculated by using ab initio HF and density functional theory calculation (B3LYP) with 6-311+G(d,p) basis set. Complete vibrational assignment and analysis of the fundamental modes of the compound were carried out using the observed FTIR and FT Raman data. The thermodynamic functions of the title compound were also performed with the aid of HF/6-311+G(d,p) and B3LYP/6-311+G(d,p) levels of theory. Simulated FTIR and FT Raman spectra for p-tosyl isocyanate showed good agreement with the observed spectra. The calculated HOMO and LUMO energies show that charge transfer occurs within the molecule. The dipole moment (μ), polarizability (α) and the hyperpolarizability (β) values of the investigated molecule have been computed using HF and B3LYP methods.