MilliporeSigma
  • Retention index database for identification of general green leaf volatiles in plants by coupled capillary gas chromatography-mass spectrometry.
Retention index database for identification of general green leaf volatiles in plants by coupled capillary gas chromatography-mass spectrometry.
Journal of chromatography. A (2000-09-29)
J Ruther
ABSTRACT
A series of ubiquitously occurring saturated and monounsaturated six-carbon aldehydes, alcohols and esters thereof is summarised as 'green leaf volatiles' (GLVs). The present study gives a comprehensive data collection of retention indices of 35 GLVs on commonly used non-polar DB-5, mid-polar DB-1701, and polar DB-Wax stationary phases. Seventeen commercially not available compounds were synthesised. Thus, the present study allows reliable identification of most known GLV in natural plant volatile samples. Applications revealed the presence of several seldom reported GLVs in headspace samples of mechanically damaged plant leaves of Carpinus betulus and Fagus sylvatica.
Product Description
Supelco
1-Hexanol, analytical standard