MilliporeSigma
Reductive generation of stable, five-membered N,N'-diamidocarbenes.
Chemical communications (Cambridge, England) (2014-03-29)
Jonathan P Moerdyk, Christopher W Bielawski
ABSTRACT
The synthesis of the first stable, five-membered N,N'-diamidocarbenes (DACs), including a differentially N-substituted derivative, was achieved via the reduction of a geminal dichloride precursor using potassium. Key differences between the reactivity of the five-membered DACs and their six-membered congeners were observed, including an ability to insert into electron-rich C-H bonds.
Sigma-Aldrich
N,N′-Di-tert-butylcarbodiimide, 99%