MilliporeSigma
  • 3-amino-4-(3-hexylphenylamino)-4-oxobutyl phosphonic acid (W146), a Selective Antagonist of Sphingosine-1-phospahte Receptor Subtype 1, Enhances AMD3100-stimulated Mobilization of Hematopoietic Stem Progenitor Cells in Animals.
3-amino-4-(3-hexylphenylamino)-4-oxobutyl phosphonic acid (W146), a Selective Antagonist of Sphingosine-1-phospahte Receptor Subtype 1, Enhances AMD3100-stimulated Mobilization of Hematopoietic Stem Progenitor Cells in Animals.
Journal of biochemical and pharmacological research (2014-11-11)
Jingjing Liu, Jiawei Zhao, Jen-Fu Lee, Allison Gartung, Hiba Jawadi, Wenliang Zhang, David Lominadze, Menq-Jer Lee
ABSTRACT
Sphingosine-1-phosphate (S1P), a serum-borne bioactive lipid, regulates various physiological functions. We observed that the S1P receptor subtype 1 (S1P1), a high affinity G-protein coupled receptor of S1P, is the major S1P receptor expressed in the Kit+/Sca-1+/Lin- (KSL) hematopoietic stem progenitor cells (HSPCs, KSL-HSPCs). In this study, we investigate function of S1P1 receptors in the regulation of HSPC mobilization in animals. Treatment with SEW2871, a specific agonist of S1P1, had no effect on KSL-HSPC mobilization. In addition, mice pretreated with SEW2871 followed by AMD3100, a well-known activator of KSL-HSPC mobilization by antagonizing the stromal-derived factor-1 (SDF-1)/C-X-C chemokine receptor type 4 (CXCR4) signaling axis, did not enhance the AMD3100-induced KSL-HSPC mobilization. In contrast, pretreatment of (R)-3-amino-4-(3-hexylphenylamino)-4-oxobutyl phosphonic acid (W146), a selective antagonist of S1P1, significantly augments AMD3100-induced KSL-HSPC mobilization into peripheral blood. The inactive enantiomer W140 was incapable of enhancing the AMD3100-induced KSL-HSPC mobilization. Moreover, treatment with selective antagonists for S1P2 and S1P3 had no effects on AMD3100-mediated KSL-HSPC mobilization. Collectively, our data suggest that S1P/S1P1 signaling regulates the SDF-1/CXCR4-mediated retention of KSL-HSPCs in bone marrow microenvironment.
Product Description
Avanti
W140, (S)-3-amino-4-(3-hexylphenylamino)-4-oxobutylphosphonic acid (TFA salt), powder