MilliporeSigma
  • Comparative profiling and comprehensive quantification of stratum corneum ceramides in humans and mice by LC/MS/MS.
Comparative profiling and comprehensive quantification of stratum corneum ceramides in humans and mice by LC/MS/MS.
Journal of lipid research (2020-04-09)
Momoko Kawana, Masatoshi Miyamoto, Yusuke Ohno, Akio Kihara
ABSTRACT
Ceramides are the predominant lipids in the stratum corneum (SC) and are crucial components for normal skin barrier function. Although the composition of various ceramide classes in the human SC has been reported, that in mice is still unknown, despite mice being widely used as animal models of skin barrier function. Here, we performed LC/MS/MS analyses using recently available ceramide class standards to measure 25 classes of free ceramides and 5 classes of protein-bound ceramides from human and mouse SC. Phytosphingosine- and 6-hydroxy sphingosine-type ceramides, which both contain an additional hydroxyl group, were abundant in the human SC (35% and 45% of total ceramides, respectively). In contrast, in mice, phytosph-ingosine- and 6-hydroxy sphingosine-type ceramides were present at ∼1% and undetectable levels, respectively, and sphingosine-type ceramides accounted for ∼90%. In humans, ceramides containing α-hydroxy FA were abundant, whereas ceramides containing β-hydroxy or ω-hydroxy FA were abundant in mice. The hydroxylated β-carbon in β-hydroxy ceramides was in the (R) configuration. Genetic knockout of β-hydroxy acyl-CoA dehydratases in HAP1 cells increased β-hydroxy ceramide levels, suggesting that β-hydroxy acyl-CoA, an FA-elongation cycle intermediate in the ER, is a substrate for β-hydroxy ceramide synthesis. We anticipate that our methods and findings will help to elucidate the role of each ceramide class in skin barrier formation and in the pathogenesis of skin disorders.
Sigma-Aldrich
DL-β-Hydroxymyristic acid, ≥98%
Avanti
CER5-2′R(d9), Avanti Research - A Croda Brand
Avanti
CER11-2′R(d9), Avanti Research - A Croda Brand 860836P, powder
Avanti
CER7-2′R,6R(d9), Avanti Research - A Croda Brand 860837P, powder
Avanti
CER10(d9), N-palmitoyl(d9) dihydrosphingosine, powder
Avanti
CER1 (d18:1/26:0/18:1(d9)), Avanti Research - A Croda Brand 860849P, powder
Avanti
CER6-2′R(d9), Avanti Research - A Croda Brand 860618P, powder