MilliporeSigma
  • DNA Intercalation Facilitates Efficient DNA-Targeted Covalent Binding of Phenanthriplatin.
DNA Intercalation Facilitates Efficient DNA-Targeted Covalent Binding of Phenanthriplatin.
Journal of the American Chemical Society (2019-01-03)
Ali A Almaqwashi, Wen Zhou, M Nabuan Naufer, Imogen A Riddell, Ömer H Yilmaz, Stephen J Lippard, Mark C Williams
ABSTRACT
Phenanthriplatin, a monofunctional anticancer agent derived from cisplatin, shows significantly more rapid DNA covalent-binding activity compared to its parent complex. To understand the underlying molecular mechanism, we used single-molecule studies with optical tweezers to probe the kinetics of DNA-phenanthriplatin binding as well as DNA binding to several control complexes. The time-dependent extensions of single λ-DNA molecules were monitored at constant applied forces and compound concentrations, followed by rinsing with a compound-free solution. DNA-phenanthriplatin association consisted of fast and reversible DNA lengthening with time constant τ ≈ 10 s, followed by slow and irreversible DNA elongation that reached equilibrium in ∼30 min. In contrast, only reversible fast DNA elongation occured for its stereoisomer  trans-phenanthriplatin, suggesting that the distinct two-rate kinetics of phenanthriplatin is sensitive to the geometric conformation of the complex. Furthermore, no DNA unwinding was observed for pyriplatin, in which the phenanthridine ligand of phenanthriplatin is replaced by the smaller pyridine molecule, indicating that the size of the aromatic group is responsible for the rapid DNA elongation. These findings suggest that the mechanism of binding of phenanthriplatin to DNA involves rapid, partial intercalation of the phenanthridine ring followed by slower substitution of the adjacent chloride ligand by, most likely, the N7 atom of a purine base. The cis isomer affords the proper stereochemistry at the metal center to facilitate essentially irreversible DNA covalent binding, a geometric advantage not afforded by trans-phenanthriplatin. This study demonstrates that reversible DNA intercalation provides a robust transition state that is efficiently converted to an irreversible DNA-Pt bound state.
Sigma-Aldrich
Phenanthriplatin, >97% (NMR)