MilliporeSigma
  • Quantification of aroma compounds in Parmigiano Reggiano cheese by a dynamic headspace gas chromatography-mass spectrometry technique and calculation of odor activity value.
Quantification of aroma compounds in Parmigiano Reggiano cheese by a dynamic headspace gas chromatography-mass spectrometry technique and calculation of odor activity value.
Journal of dairy science (2003-04-22)
Michael Qian, G A Reineccius
ABSTRACT
Potentially important aroma compounds in Parmigiano Reggiano cheese were quantified. Free fatty acids were isolated with ion-exchange chromatography and quantified by gas chromatography. Neutral aroma compounds were quantified with a purge-trap/gas chromatography-mass spectrometry with selective mass ion technique. Odor activity values were calculated based on sensory thresholds reported in literature. The calculated odor activity values suggest that 3-methylbutanal, 2-methylpropanal, 2-methylbutanal, dimethyl trisulfide, diacetyl, methional, phenylacetaldehyde, ethyl butanoate, ethyl hexanoate, ethyl octanoate, acetic, butanoic, hexanoic, and octanoic acids are the most important aroma contributors to Parmigiano Reggiano cheese.
Product Description
Supelco
2-Heptanone, analytical standard