MilliporeSigma
  • Discovery of novel (1S)-(-)-verbenone derivatives with anti-oxidant and anti-ischemic effects.
Discovery of novel (1S)-(-)-verbenone derivatives with anti-oxidant and anti-ischemic effects.
Bioorganic & medicinal chemistry letters (2013-08-21)
Chung Ju, Sumi Song, Sunyoung Hwang, Chorong Kim, Minkyoung Kim, Jail Gu, Yu-Kyoung Oh, Kyeong Lee, Jinsun Kwon, Kiho Lee, Won-Ki Kim, Yongseok Choi
ABSTRACT
A series of novel (1S)-(-)-verbenone derivatives was synthesized bearing a 4-styryl scaffold. The synthesized compounds were tested for their anti-oxidant, anti-excitotoxic, and anti-ischemic activities. These derivatives significantly reduced oxygen-glucose deprivation-induced neuronal injury and N-methyl-D-aspartic acid-evoked excitotoxicity in cortical neurons. Furthermore, compound 3f was identified as a potent anti-ischemic agent in an in vitro ischemic model, potentially due to the inhibition of N-methyl-D-aspartic acid-evoked excitotoxicity and oxidative/nitrosative stress.
Sigma-Aldrich
(1S)-(−)-Verbenone, 94%