MilliporeSigma
  • Overexpression of aprE2, a fibrinolytic enzyme gene from Bacillus subtilis CH3-5, in Escherichia coli and the properties of AprE2.
Overexpression of aprE2, a fibrinolytic enzyme gene from Bacillus subtilis CH3-5, in Escherichia coli and the properties of AprE2.
Journal of microbiology and biotechnology (2014-04-20)
Seon-Ju Jeong, Kye Man Cho, Chang Kwon Lee, Gyoung Min Kim, Jung-Hye Shin, Jong Sang Kim, Jeong Hwan Kim
ABSTRACT
The aprE2 gene with its prosequence from Bacillus subtilis CH3-5 was overexpressed in Escherichia coli BL21(DE3) by using plasmid pET26b(+). After IPTG induction, active and mature AprE2 was produced when cells were grown at 20°C, whereas inactive and insoluble enzyme was produced in a large amount when cells were grown at 37°C. The insoluble fraction was resuspended with 6 M guanidine-HCl and dialyzed against 2 M Tris-HCl (pH 7.0) or 0.5 M sodium acetate (pH 7.0) buffer. Then active AprE2 was regenerated and purified by a Ni-NTA column. Purified AprE2 from the soluble fraction had a specific activity of 1,069.4 ± 42.4 U/mg protein, higher than that from the renatured insoluble fraction. However, more active AprE2 was obtained by renaturation of the insoluble fraction. AprE2 was most stable at pH 7 and 40°C, respectively. The fibrinolytic activity of AprE2 was inhibited by PMSF, but not by EDTA and metal ions. AprE2 degraded Aα and Bβ chains of fibrinogen quickly, but not the γ-chain. AprE2 exhibited the highest specificity for N-succinyl-Ala-Ala-Pro-Phe-pNA. The Km and kcat/Km of AprE2 was 0.56 mM and 3.10 × 10(4) S(-1) M(-1), respectively.
Sigma-Aldrich
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, protease substrate