MilliporeSigma
  • Metabolic Stability Assessment of Larotrectinib Using Liquid Chromatography Tandem Mass Spectrometry.
Metabolic Stability Assessment of Larotrectinib Using Liquid Chromatography Tandem Mass Spectrometry.
Drug design, development and therapy (2020-02-06)
Mohamed W Attwa, Adnan A Kadi, Hany W Darwish
ABSTRACT
Larotrectinib (VITRAKVI) is an orally potent tropomyosin receptor kinase (Trk) inhibitor that acts by competitive inhibition of all corresponding receptor kinases. It demonstrated a marked response rate (75%) and robust anticancer activity in Trk fusion-positive patients. This response is independent of cancer type, age and gender. In this study, an efficient and accurate LC-MS/MS analytical method was developed for Larotrectinib (LRB) quantification in addition to evaluation of its metabolic stability. LRB and lapatinib (LTP) (which is chosen as an internal standard; IS) were eluted utilizing an isocratic mobile phase with a reversed phase elution system (C18 column). The linearity range of the established method was 5-500 ng/mL (r2 ≥ 0.9999) in the human liver microsomes (HLMs) matrix. Various parameters were calculated to validate the method sensitivity (limit of quantification was 5 ng/mL) and reproducibility (inter and intra-day accuracy and precision were below 3% in all samples) of our methodology. For evaluation of LRB metabolic stability in HLMs matrix, in vitro half-life (48.8 min) and intrinsic clearance (14.19 µL/min/mg) were computed. Accordingly, we can conclude that LRB is a moderate extraction ratio drug when compared with other tyrosine kinase inhibitors (TKIs). According to our knowledge, the discussed procedure in this study is the first LC-MS/MS analytical method for evaluating LRB metabolic stability.
Sigma-Aldrich
Microsomes from Liver, Pooled, from human male