MilliporeSigma
  • Relationship between digestive enzymes and food habit of Lutzomyia longipalpis (Diptera: Psychodidae) larvae: Characterization of carbohydrases and digestion of microorganisms.
Relationship between digestive enzymes and food habit of Lutzomyia longipalpis (Diptera: Psychodidae) larvae: Characterization of carbohydrases and digestion of microorganisms.
Journal of insect physiology (2012-06-12)
C S Moraes, S A Lucena, B H S Moreira, R P Brazil, N F Gontijo, F A Genta
ABSTRACT
The sandfly Lutzomyia longipalpis (Lutz and Neiva, 1912) is the main vector of American Visceral Leishmaniasis. In spite of its medical importance and several studies concerning adult digestive physiology, biochemistry and molecular biology, very few studies have been carried out to elucidate the digestion in sandfly larvae. Even the breeding sites and food sources of these animals in the field are largely uncharacterized. In this paper, we describe and characterize several carbohydrases from the gut of L. longipalpis larvae, and show that they are probably not acquired from food. The enzyme profile of this insect is consistent with the digestion of fungal and bacterial cells, which were proved to be ingested by larvae under laboratory conditions. In this respect, sandfly larvae might have a detritivore habit in nature, being able to exploit microorganisms usually encountered in the detritus as a food source.
Sigma-Aldrich
4-Methylumbelliferyl β-D-mannopyranoside, ≥98% (TLC)