MilliporeSigma
  • Total Synthesis of Integrastatin B Enabled by a Benzofuran Oxidative Dearomatization Cascade.
Total Synthesis of Integrastatin B Enabled by a Benzofuran Oxidative Dearomatization Cascade.
Organic letters (2016-03-08)
Atul A More, Chepuri V Ramana
ABSTRACT
The first total synthesis of integrastatin B, a potent HIV-1 integrase inhibitor, has been accomplished in seven steps with a 17.9% overall yield employing easily accessible starting compounds. The Oxone-mediated oxidative benzofuran dearomatization cascade has been employed as the key skeletal construct to forge the central tetracyclic nucleus.