MilliporeSigma
  • Immunoglobulin G1 Antibodies Against Phosphorylcholine Are Associated With Protection in Systemic Lupus Erythematosus and Atherosclerosis: Potential Underlying Mechanisms.
Immunoglobulin G1 Antibodies Against Phosphorylcholine Are Associated With Protection in Systemic Lupus Erythematosus and Atherosclerosis: Potential Underlying Mechanisms.
ACR open rheumatology (2020-05-12)
D Thiagarajan, R Fiskesund, A Frostegård, J Steen, M Rahman, M Vikström, S Lundström, J Frostegård
ABSTRACT
Immunoglobulin M antibodies against phosphorylcholine (anti-PCs) may be protective in atherosclerosis, cardiovascular disease (CVD), and systemic lupus erythematosus (SLE). We study immunoglobulin G1 (IgG1) and immunoglobulin G2 (IgG2) anti-PCs, with a focus on atherosclerosis and SLE. We determined anti-PCs by using the enzyme-linked immunosorbent assay in 116 patients with SLE and 110 age- and sex-matched controls. For functional studies, we used three in-house-generated, fully human monoclonal IgG1 anti-PCs (A01, D05, and E01). Apoptosis was induced in Jurkat T cells and preincubated with A01, D05, E01, or IgG1 isotype control, and effects on efferocytosis by human macrophages were studied. Anti-PC peptide/protein characterization was determined using a proteomics de novo sequencing approach. IgG1, but not IgG2, anti-PC levels were higher among patients with SLE (P = 0.02). IgG1 anti-PCs were negatively associated with Systemic Lupus International Collaborating Clinics (SLICC) damage index and Systemic Lupus Erythematosus Disease Activity Index (SLEDAI) scores (odds ratio [OR]: 2.978 [confidence interval (CI): 0.876-10.098] and OR: 5.108 [CI 1.3-20.067], respectively) and negatively associated with CVD, atherosclerotic plaques, and echolucent plaques (potentially vulnerable plaques), but the association for the two former was not significant after controlling for confounders. D05 had a maximum effect on macrophage efferocytosis efficiency, followed by A01 and E01. The monoclonal antibodies showed differential binding specificity to PC and PC-associated neoepitopes. A peptide analysis showed a difference in the complementarity-determining region 3 of the three IgG1 anti-PC clones that are crucial for recognition of PC on apoptotic cell surfaces and other neoepitopes. IgG1 anti-PCs are negatively associated with disease activity and disease damage in SLE, but the negative association with CVD is also dependent on confounding risk factors. One potential underlying mechanism could be increased clearance of dead cells.
Sigma-Aldrich
Anti-Human IgG1−Biotin antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG2 antibody produced in mouse, clone HP-6014, ascites fluid
Avanti
POVPC, Avanti Research - A Croda Brand 870606P, powder