MilliporeSigma
  • Chemical structures of hydrazine-treated graphene oxide and generation of aromatic nitrogen doping.
Chemical structures of hydrazine-treated graphene oxide and generation of aromatic nitrogen doping.
Nature communications (2012-01-26)
Sungjin Park, Yichen Hu, Jin Ok Hwang, Eui-Sup Lee, Leah B Casabianca, Weiwei Cai, Jeffrey R Potts, Hyung-Wook Ha, Shanshan Chen, Junghoon Oh, Sang Ouk Kim, Yong-Hyun Kim, Yoshitaka Ishii, Rodney S Ruoff
ABSTRACT
Chemically modified graphene platelets, produced via graphene oxide, show great promise in a variety of applications due to their electrical, thermal, barrier and mechanical properties. Understanding the chemical structures of chemically modified graphene platelets will aid in the understanding of their physical properties and facilitate development of chemically modified graphene platelet chemistry. Here we use (13)C and (15)N solid-state nuclear magnetic resonance spectroscopy and X-ray photoelectron spectroscopy to study the chemical structure of (15)N-labelled hydrazine-treated (13)C-labelled graphite oxide and unlabelled hydrazine-treated graphene oxide, respectively. These experiments suggest that hydrazine treatment of graphene oxide causes insertion of an aromatic N(2) moiety in a five-membered ring at the platelet edges and also restores graphitic networks on the basal planes. Furthermore, density-functional theory calculations support the formation of such N(2) structures at the edges and help to elucidate the influence of the aromatic N(2) moieties on the electronic structure of chemically modified graphene platelets.
Sigma-Aldrich
3D Printable Graphene Oxide Ink, avg. no. of layers, 1