MilliporeSigma
  • Long-term use of fluoxetine accelerates bone loss through the disruption of sphingolipids metabolism in bone marrow adipose tissue.
Long-term use of fluoxetine accelerates bone loss through the disruption of sphingolipids metabolism in bone marrow adipose tissue.
Translational psychiatry (2020-05-14)
Huili Zhang, Kefeng Li, Yanna Zhao, Yilan Zhang, Jiawen Sun, Shihong Li, Guangwu Lin
ABSTRACT
Fluoxetine is a commonly prescribed antidepressant, and the mechanisms of increased bone fragility with its long-term use remain largely unknown. Here, we show that long-term administration of fluoxetine induces the disruption of sphingolipids metabolism in bone marrow adipose tissue (BMAT)through the inhibition of acid sphingomyelinase (ASM). Similarly, a significant reduction of the bone volume was observed in mice with ASM knockout (Smpd1-/-). In detail, inhibition of ASM by fluoxetine reduces the sphingosine-1-phosphate (S1P) level in bone marrow adipocytes, leading to the increase of receptor activator of nuclear factor-kappa-Β ligand (RANKL) secretion, a key regulator for the activation of osteoclastogenesis and bone loss, through the upregulation of cyclooxygenase-2 and its enzymatic product prostaglandin E2 (COX-2/PGE2). In contrast, overexpression of ASM by cisplatin normalizes fluoxetine-induced RANKL overproduction. Furthermore, we conducted a clinical trial with L-serine, a precursor of sphingolipids biosynthesis. The results show that oral supplementation of L-serine (250 mg//kg/d) prevents the acceleration of bone loss caused by long-term fluoxetine (12 months) in postmenopausal women with major depressive disorder (mean total hip bone mineral density reduction: -2.0% vs -1.1%, P = 0.006). Our study provides new insights and potential treatment strategy on the bone loss caused by long-term use of fluoxetine.
Product Description
Avanti
sphingosine-1-phosphate-d7, Avanti Research - A Croda Brand 860659P, powder
Avanti
17:0 SM (d18:1/17:0), Avanti Research - A Croda Brand 860585P, powder