MilliporeSigma
  • Short-Term Effect of the Inclusion of Silage Artichoke By-Products in Diets of Dairy Goats on Milk Quality.
Short-Term Effect of the Inclusion of Silage Artichoke By-Products in Diets of Dairy Goats on Milk Quality.
Animals : an open access journal from MDPI (2020-02-27)
Paula Monllor, Gema Romero, Esther Sendra, Alberto Stanislao Atzori, José Ramón Díaz
ABSTRACT
Artichoke by-products represent a high amount of waste whose removal entails several costs. Moreover, feed is the main cost in a farm. So, including these by-products in ruminant diets would lower feed costs. Two experiments were conducted to evaluate the effect of two levels of inclusion, 12.5% and 25.0%, of two silages of artichoke by-products (artichoke bracts, AB and artichoke plant, AP) in the diet of goats on the milk yield, composition and quality and on the metabolic profile of the animals. AB presented the lowest blood urea content and there were no differences in milk yield in the two experiments. However, with 25.0% of silage by-product in the diet, a higher fat content was observed in AB and of protein in AP, as well as this treatment showing a slightly higher Se content. Regarding the milk lipid profile, milk from 12.5% of AP treatment presented a higher PUFA content. In conclusion, the use of silage artichoke by-products in dairy goat diets does not jeopardise milk yield and quality and health status of animals and, from a nutritional point of view for human health, a slightly better mineral and lipid profile is observed in milk from AP treatments.