MilliporeSigma
  • MHC-E-Restricted CD8+ T Cells Target Hepatitis B Virus-Infected Human Hepatocytes.
MHC-E-Restricted CD8+ T Cells Target Hepatitis B Virus-Infected Human Hepatocytes.
Journal of immunology (Baltimore, Md. : 1950) (2020-03-13)
Benjamin J Burwitz, Patrick K Hashiguchi, Mandana Mansouri, Christine Meyer, Roxanne M Gilbride, Sreya Biswas, Jennie L Womack, Jason S Reed, Helen L Wu, Michael K Axthelm, Scott G Hansen, Louis J Picker, Klaus Früh, Jonah B Sacha
ABSTRACT
Currently 247 million people are living with chronic hepatitis B virus infection (CHB), and the development of novel curative treatments is urgently needed. Immunotherapy is an attractive approach to treat CHB, yet therapeutic approaches to augment the endogenous hepatitis B virus (HBV)-specific T cell response in CHB patients have demonstrated little success. In this study, we show that strain 68-1 rhesus macaque (RM) CMV vaccine vectors expressing HBV Ags engender HBV-specific CD8+ T cells unconventionally restricted by MHC class II and the nonclassical MHC-E molecule in RM. Surface staining of human donor and RM primary hepatocytes (PH) ex vivo revealed the majority of PH expressed MHC-E but not MHC class II. HBV-specific, MHC-E-restricted CD8+ T cells from RM vaccinated with RM CMV vaccine vectors expressing HBV Ags recognized HBV-infected PH from both human donor and RM. These results provide proof-of-concept that MHC-E-restricted CD8+ T cells could be harnessed for the treatment of CHB, either through therapeutic vaccination or adoptive immunotherapy.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, for molecular biology, 6,000