MilliporeSigma
  • Fate and Bioaccessibility of Iodine in Food Prepared from Agronomically Biofortified Wheat and Rice and Impact of Cofertilization with Zinc and Selenium.
Fate and Bioaccessibility of Iodine in Food Prepared from Agronomically Biofortified Wheat and Rice and Impact of Cofertilization with Zinc and Selenium.
Journal of agricultural and food chemistry (2020-01-17)
Ismail Cakmak, Massimo Marzorati, Pieter Van den Abbeele, Katja Hora, Harmen Tjalling Holwerda, Mustafa Atilla Yazici, Erdinc Savasli, Joachim Neri, Gijs Du Laing
ABSTRACT
Enrichment of food crops with iodine is an option to alleviate dietary deficiencies. Therefore, foliar iodine fertilizer was applied on wheat and rice, in the presence and absence of the other micronutrients zinc and selenium. This treatment increased the concentration of iodine, as well as zinc and selenium, in the staple grains. Subsequently, potential iodine losses during preparation of foodstuffs with the enriched grains were studied. Oven-heating did not affect the iodine content in bread. Extraction of bran from flour lowered the iodine in white bread compared to wholegrain bread, but it was still markedly higher compared to the control. During subsequent in vitro gastrointestinal digestion, a higher percentage of iodine was released from foods based on extracted flour (82-92%) compared to wholegrain foods (50-76%). The foliar fertilization of wheat was found to be adequate to alleviate iodine deficiency in a population with a moderate to high intake of bread.
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
α-Amylase from hog pancreas, powder, ~50 U/mg