MilliporeSigma
  • The stress-responsive gene GDPGP1/mcp-1 regulates neuronal glycogen metabolism and survival.
The stress-responsive gene GDPGP1/mcp-1 regulates neuronal glycogen metabolism and survival.
The Journal of cell biology (2020-01-23)
Alexander Schulz, Yuichi Sekine, Motunrayo J Oyeyemi, Alexander J Abrams, Manasa Basavaraju, Sung Min Han, Marco Groth, Helen Morrison, Stephen M Strittmatter, Marc Hammarlund
ABSTRACT
Maladaptive responses to stress might play a role in the sensitivity of neurons to stress. To identify novel cellular responses to stress, we performed transcriptional analysis in acutely stressed mouse neurons, followed by functional characterization in Caenorhabditis elegans. In both contexts, we found that the gene GDPGP1/mcp-1 is down-regulated by a variety of stresses. Functionally, the enzyme GDPGP1/mcp-1 protects against stress. Knockdown of GDPGP1 in mouse neurons leads to widespread neuronal cell death. Loss of mcp-1, the single homologue of GDPGP1 in C. elegans, leads to increased degeneration of GABA neurons as well as reduced survival of animals following environmental stress. Overexpression of mcp-1 in neurons enhances survival under hypoxia and protects against neurodegeneration in a tauopathy model. GDPGP1/mcp-1 regulates neuronal glycogen levels, indicating a key role for this metabolite in neuronal stress resistance. Together, our data indicate that down-regulation of GDPGP1/mcp-1 and consequent loss of neuronal glycogen is a maladaptive response that limits neuronal stress resistance and reduces survival.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)