MilliporeSigma
  • Differential regulation of breast cancer bone metastasis by PARP1 and PARP2.
Differential regulation of breast cancer bone metastasis by PARP1 and PARP2.
Nature communications (2020-03-30)
Hao Zuo, Dengbao Yang, Qiwen Yang, Haidong Tang, Yang-Xin Fu, Yihong Wan
ABSTRACT
PARP1 and PARP2 dual inhibitors, such as olaparib, have been recently FDA approved for the treatment of advanced breast and ovarian cancers. However, their effects on bone mass and bone metastasis are unknown. Here we show that olaparib increases breast cancer bone metastasis through PARP2, but not PARP1, specifically in the myeloid lineage, but not in the cancer cells. Olaparib treatment or PARP1/2 deletion promotes osteoclast differentiation and bone loss. Intriguingly, myeloid deletion of PARP2, but not PARP1, increases the population of immature myeloid cells in bone marrow, and impairs the expression of chemokines such as CCL3 through enhancing the transcriptional repression by β-catenin. Compromised CCL3 production in turn creates an immune-suppressive milieu by altering T cell subpopulations. Our findings warrant careful examination of current PARP inhibitors on bone metastasis and bone loss, and suggest cotreatment with CCL3, β-catenin inhibitors, anti-RANKL or bisphosphonates as potential combination therapy for PARP inhibitors.
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PARP2
Sigma-Aldrich
MISSION® esiRNA, targeting human PARP1
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes