MilliporeSigma
  • Continuous Radio Amplification by Stimulated Emission of Radiation using Parahydrogen Induced Polarization (PHIP-RASER) at 14 Tesla.
Continuous Radio Amplification by Stimulated Emission of Radiation using Parahydrogen Induced Polarization (PHIP-RASER) at 14 Tesla.
Chemphyschem : a European journal of chemical physics and physical chemistry (2020-01-04)
Andrey N Pravdivtsev, Frank D Sönnichsen, Jan-Bernd Hövener
ABSTRACT
Nuclear Magnetic Resonance (NMR) is an intriguing quantum-mechanical effect that is used for routine medical diagnostics and chemical analysis alike. Numerous advancements have contributed to the success of the technique, including hyperpolarized contrast agents that enable real-time imaging of metabolism in vivo. Herein, we report the finding of an NMR radio amplification by stimulated emission of radiation (RASER), which continuously emits 1 H NMR signal for more than 10 min. Using parahydrogen induced hyperpolarization (PHIP) with 50 % para-hydrogen, we demonstrated the effect at 600 MHz but expect that it is functional across a wide range of frequencies, e.g. 101 -103  MHz. PHIP-RASER occurs spontaneously or can be triggered with a standard NMR excitation. Full chemical shift resolution was maintained, and a linewidth of 0.6 ppb was achieved. The effect was reproduced by simulations using a weakly coupled, two spin- 1 / 2 system. All devices used were standard issue, such that the effect can be reproduced by any NMR lab worldwide with access to liquid nitrogen for producing parahydrogen.
Sigma-Aldrich
Methyl propiolate, 99%
Sigma-Aldrich
Ethyl phenylpropiolate, 98%
Sigma-Aldrich
[1,4-Bis(diphenylphosphino)butane](1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98%