MilliporeSigma
  • COUP-TFII Knock-down Promotes Proliferation and Invasion in Colorectal Cancer Cells via Activation of Akt Pathway and Up-regulation of FOXC1.
COUP-TFII Knock-down Promotes Proliferation and Invasion in Colorectal Cancer Cells via Activation of Akt Pathway and Up-regulation of FOXC1.
Anticancer research (2020-01-02)
Seong-Hoon Yun, Sang-Heum Han, Joo-In Park
ABSTRACT
The chicken ovalbumin upstream promoter-transcription factor II (COUP-TFII) regulates cancer cell proliferation and invasion via complex molecular mechanisms. We aimed to investigate whether COUP-TFII modulates proliferation and invasion of the colorectal adenocarcinoma cell line HT-29. HT-29 cells were stably tranfected with COUP-TFII shRNA plasmid to knock-down COUP-TFII (COUP-TFII shRNA-HT-29 cells). Cell proliferation, colony formation assay, invasion assay, microarray assays and western blot analyses were performed. Cell proliferation and invasion were significantly enhanced in COUP-TFII shRNA-HT-29 cells. The protein levels of forkhead box C1 (FOXC1), p-Akt, p-glycogen synthase kinase-3β (p-GSK-3β), and β-catenin, which are known to be involved in cell proliferation and invasion, were significantly increased in COUP-TFII shRNA-HT-29 cells. Akt inhibitor IV and dominant negative (DN)-Akt expression vector transfection reversed the increased proliferation and invasion, which was accompanied by decreased protein levels of p-Akt, p-GSK-3β, β-catenin and FOXC1. COUP-TFII knock-down promoted proliferation and invasion via activation of Akt/GSK-3β/β-catenin and up-regulation of FOXC1. Further studies on the molecular mechanism of interaction between β-catenin and FOXC1 expression may reveal novel target molecules for metastatic colorectal cancer therapy.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Akt Inhibitor IV, InSolution, ≥98%
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture