MilliporeSigma
  • Amifostine remediates the degenerative effects of radiation on the mineralization capacity of the murine mandible.
Amifostine remediates the degenerative effects of radiation on the mineralization capacity of the murine mandible.
Plastic and reconstructive surgery (2012-03-30)
Catherine N Tchanque-Fossuo, Alexis Donneys, Sagar S Deshpande, Noah S Nelson, Matt J Boguslawski, K Kelly Gallagher, Deniz Sarhaddi, Behdod Poushanchi, Steven A Goldstein, Steven R Buchman
ABSTRACT
Radiotherapy, a cornerstone of head and neck cancer treatment, causes substantial morbidity to normal adjoining bone. The authors assessed the radioprotective effect of amifostine therapy on the mineralization of the mandible using micro-computed tomography. They hypothesized that amifostine would safeguard the mandible from radiation-induced disruption of the mineralization process and the associated failure of new bone creation. Male Sprague-Dawley rats were randomized into three groups: control (n = 8), radiation therapy (n = 5), and amifostine (n = 8). Animals in the radiation therapy and amifostine groups underwent human bioequivalent radiation of 70 Gy in five fractions to the left hemimandible. Fifty-six days after irradiation, the hemimandibles were harvested for radiomorphometric analyses. Amifostine-treated animals exhibited less alopecia, mucositis, and weight loss in addition to increased cortical density in comparison with those treated with radiation therapy. Bone and tissue mineral densities showed statistically significant improvement in amifostine versus radiation therapy, and no difference was observed between amifostine and control groups. Detailed micro-computed tomographic analysis further demonstrated significant differences in the mineralization profile when comparing radiation therapy and amifostine. Amifostine maintained regions of lower mineralization consistent with the preservation of normal remodeling. The authors have successfully demonstrated the ability of amifostine pretreatment to protect the natural mineralization profile of bone. This reflects the capacity of amifostine prophylaxis to safeguard the normal surrounding mandible from the impediments of collateral damage imposed by irradiation. Further study can correlate these findings with the potential use of amifostine to prevent the devastating associated morbidities of radiotherapy such as pathologic fractures and osteoradionecrosis.
Sigma-Aldrich
Amifostine trihydrate, ≥97% (TLC), powder