MilliporeSigma
  • The use of autologous platelet-rich plasma (PRP) versus no intervention in women with low ovarian reserve undergoing fertility treatment: a non-randomized interventional study.
The use of autologous platelet-rich plasma (PRP) versus no intervention in women with low ovarian reserve undergoing fertility treatment: a non-randomized interventional study.
Journal of assisted reproduction and genetics (2020-02-08)
P Melo, C Navarro, C Jones, K Coward, L Coleman
ABSTRACT
To investigate the impact of a 3-month course of intracortical injections of autologous platelet-rich plasma (PRP) upon ovarian reserve markers versus no intervention in women with low ovarian reserve prior to undergoing assisted reproductive technology (ART). Prospective controlled, non-randomized comparative study conducted in a private fertility clinic, in Venezuela. Women with abnormal ovarian reserve markers (FSH, AMH and AFC) who declined oocyte donation were allocated to one of the following groups according to patient choice: monthly intracortical ovarian PRP injections for three cycles, or no intervention. Primary outcomes were the change in FSH, AMH and AFC pre- and post-treatment. Secondary outcomes included the number of oocytes collected and fertilized, biochemical/clinical pregnancy rates and miscarriage and live birth rates. Eighty-three women were included, of which 46 received PRP treatment and 37 underwent no intervention. Overall median age was 41 years (IQR 39-44). There were no demographic differences between the study groups. At the 3-month follow-up, women treated with PRP experienced a significant improvement in FSH, AMH and AFC, whereas there was no change in the control group. Furthermore, overall rates of biochemical (26.1% versus 5.4%, P = 0.02) and clinical pregnancy (23.9% versus 5.4%, P = 0.03) were higher in the PRP group, while there was no difference in the rates of first trimester miscarriage and live birth between groups. PRP injections are effective and safe to improve markers of low ovarian reserve prior to ART, although further evidence is required to evaluate the impact of PRP on pregnancy outcomes.
Sigma-Aldrich
Cetrorelix acetate, ≥98% (HPLC)
Sigma-Aldrich
Human Siglec-5 ELISA Kit, for serum, plasma, cell culture supernatants and urine