MilliporeSigma
  • Multiple draft tubes airlift loop membrane bioreactor as an efficient system for acidic 7-amino cephalosporanic acid (7-ACA) wastewater treatment.
Multiple draft tubes airlift loop membrane bioreactor as an efficient system for acidic 7-amino cephalosporanic acid (7-ACA) wastewater treatment.
Bioresource technology (2020-02-24)
Dongxue Hu, Yuanyi Zhao, Hongcheng Wang, Hongchao Min, Yubo Cui, Kongyan Luo, Lufeng Zhang, Wenyu Liu, Ying Zhang
ABSTRACT
In this study, a lab-scale multiple draft tubes airlift loop membrane bioreactor (Mt-ALMBR) was used for treating acidic 7-amino cephalosporanic acid (7-ACA) wastewater under different pHs (3.54-6.20) and hydraulic retention time (HRT) (48 h, 36 h, 24 h and 16 h). During about 200 days operation, under HRT of 48 h and pH condition about 6.0, the optimum average COD and BOD5 removal rates were reach to 84.4 ± 2.1% and 94.9 ± 0.8%, and the highest 7-ACA removal rate also observed as 77.6%. Biodegradation, membrane rejection, hydrolysis and sludge adsorption were the four main pathways of 7-ACA removal. With the increase of pH, biodegradation, membrane rejection and hydrolysis had significant positive impacts on 7-ACA removal, while adsorption had a negative impact. Moreover, mathematical models for 7-ACA removal rate and pH were calculated to guide the operation of Mt-ALMBR. Biodegradation was the main pathway to remove 7-ACA when pH was >4.17.
Sigma-Aldrich
7-Aminocephalosporanic acid, 98%