MilliporeSigma
  • Simultaneous tracers and a unified model of positional and mass isotopomers for quantification of metabolic flux in liver.
Simultaneous tracers and a unified model of positional and mass isotopomers for quantification of metabolic flux in liver.
Metabolic engineering (2020-01-01)
Stanislaw Deja, Xiaorong Fu, Justin A Fletcher, Blanka Kucejova, Jeffrey D Browning, Jamey D Young, Shawn C Burgess
ABSTRACT
Computational models based on the metabolism of stable isotope tracers can yield valuable insight into the metabolic basis of disease. The complexity of these models is limited by the number of tracers and the ability to characterize tracer labeling in downstream metabolites. NMR spectroscopy is ideal for multiple tracer experiments since it precisely detects the position of tracer nuclei in molecules, but it lacks sensitivity for detecting low-concentration metabolites. GC-MS detects stable isotope mass enrichment in low-concentration metabolites, but lacks nuclei and positional specificity. We performed liver perfusions and in vivo infusions of 2H and 13C tracers, yielding complex glucose isotopomers that were assigned by NMR and fit to a newly developed metabolic model. Fluxes regressed from 2H and 13C NMR positional isotopomer enrichments served to validate GC-MS-based flux estimates obtained from the same experimental samples. NMR-derived fluxes were largely recapitulated by modeling the mass isotopomer distributions of six glucose fragment ions measured by GC-MS. Modest differences related to limited fragmentation coverage of glucose C1-C3 were identified, but fluxes such as gluconeogenesis, glycogenolysis, cataplerosis and TCA cycle flux were tightly correlated between the methods. Most importantly, modeling of GC-MS data could assign fluxes in primary mouse hepatocytes, an experiment that is impractical by 2H or 13C NMR.
Product Description
Supelco
Discovery® DSC-18 SPE Tube, bed wt. 500 mg, volume 3 mL, pk of 54