MilliporeSigma
  • A human embryonic stem cell reporter line for monitoring chemical-induced cardiotoxicity.
A human embryonic stem cell reporter line for monitoring chemical-induced cardiotoxicity.
Cardiovascular research (2019-06-08)
Su-Yi Tsai, Zaniar Ghazizadeh, Hou-Jun Wang, Sadaf Amin, Francis A Ortega, Zohreh Sadat Badieyan, Zi-Ting Hsu, Miriam Gordillo, Ritu Kumar, David J Christini, Todd Evans, Shuibing Chen
ABSTRACT
Human embryonic stem cells (hESCs) can be used to generate scalable numbers of cardiomyocytes (CMs) for studying cardiac biology, disease modelling, drug screens, and potentially for regenerative therapies. A fluorescence-based reporter line will significantly enhance our capacities to visualize the derivation, survival, and function of hESC-derived CMs. Our goal was to develop a reporter cell line for real-time monitoring of live hESC-derived CMs. We used CRISPR/Cas9 to knock a mCherry reporter gene into the MYH6 locus of hESC lines, H1 and H9, enabling real-time monitoring of the generation of CMs. MYH6:mCherry+ cells express atrial or ventricular markers and display a range of cardiomyocyte action potential morphologies. At 20 days of differentiation, MYH6:mCherry+ cells show features characteristic of human CMs and can be used successfully to monitor drug-induced cardiotoxicity and oleic acid-induced cardiac arrhythmia. We created two MYH6:mCherry hESC reporter lines and documented the application of these lines for disease modelling relevant to cardiomyocyte biology.
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Oleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutralization Solution for Blood, sufficient for 100 reactions, sufficient for 1000 reactions, for molecular biology
Sigma-Aldrich
Lysis solution for blood, sufficient for 100 reactions, for molecular biology