MilliporeSigma
  • Liver Activation of Hepatocellular Nuclear Factor-4α by Small Activating RNA Rescues Dyslipidemia and Improves Metabolic Profile.
Liver Activation of Hepatocellular Nuclear Factor-4α by Small Activating RNA Rescues Dyslipidemia and Improves Metabolic Profile.
Molecular therapy. Nucleic acids (2019-12-27)
Kai-Wen Huang, Vikash Reebye, Katherine Czysz, Simona Ciriello, Stephanie Dorman, Isabella Reccia, Hong-Shiee Lai, Ling Peng, Nikos Kostomitsopoulos, Joanna Nicholls, Robert S Habib, Donald A Tomalia, Pål Sætrom, Edmund Wilkes, Pedro Cutillas, John J Rossi, Nagy A Habib
ABSTRACT
Non-alcoholic fatty liver disease (NAFLD) culminates in insulin resistance and metabolic syndrome. Because there are no approved pharmacological treatment agents for non-alcoholic steatohepatitis (NASH) and NAFLD, different signaling pathways are under investigation for drug development with the focus on metabolic pathways. Hepatocyte nuclear factor 4-alpha (HNF4A) is at the center of a complex transcriptional network where its disruption is directly linked to glucose and lipid metabolism. Resetting HNF4A expression in NAFLD is therefore crucial for re-establishing normal liver function. Here, small activating RNA (saRNA) specific for upregulating HNF4A was injected into rats fed a high-fat diet for 16 weeks. Intravenous delivery was carried out using 5-(G5)-triethanolamine-core polyamidoamine (PAMAM) dendrimers. We observed a significant reduction in liver triglyceride, increased high-density lipoprotein/low-density lipoprotein (HDL/LDL) ratio, and decreased white adipose tissue/body weight ratio, all parameters to suggest that HNF4A-saRNA treatment induced a favorable metabolic profile. Proteomic analysis showed significant regulation of genes involved in sphingolipid metabolism, fatty acid β-oxidation, ketogenesis, detoxification of reactive oxygen species, and lipid transport. We demonstrate that HNF4A activation by oligonucleotide therapy may represent a novel single agent for the treatment of NAFLD and insulin resistance.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Lard oil